Nalgene® Filter Storage Bottle Receivers, Sterile, Thermo Scientific
# de catalogue 28199-189
Fournisseur: Thermo Fisher Scientific
Spécifications
- Capacity:500 ml
- Supplier No.:455-0500
Spécifications
A propos de cet article
These receiver storage bottles feature an ergonomic design with tapered sides and grips for ease of handling.
- Integral no-drip lip facilitates pouring
- Easy-to-read graduations on two sides
- Non-cytotoxic, non-pyrogenic, radiation sterilized
Storage bottles have a 45 mm Ø closure and are designed for use with Nalgene® Rapid-Flow™ series bottle-top filters and filter units. A 1¹/₂ -turn threaded screw closure guarantees a leakproof seal and reduces pH shift of stored media. The storage bottles have at least 20% extra head space for the addition of serum or other additives to the filtrate.
Customers Who Bought This Also Bought