Spécifications
- Description:B2 System with Buffer Exchange Ports
- Cat. no.:27372-258
- L×W×H:24.5 x 18 x 8 cm (931/32 x 7 x 35/32")
- Buffer Volume:800 ml
- Gel Dimensions:12 x 14 cm
- Supplier No.:B2-BP
Spécifications
À propos de cet article
Mini-Gel systems have a small footprint and can run 5–34 samples (for 400 and 600mL systems) or 8–48 samples (for 800mL systems) on one gel
Leak-resistant, gasketed ultraviolet transmission trays allow tape-free casting in the buffer chamber. The trays have two comb positions and allow DNA bands to be viewed on a transilluminator without removing the gel. Optional multiload UVT trays have 12 slots for multiple sample-running, and allow samples to be loaded directly from a 96-well plate into an agarose gel. This design is based on the fixed spacing required to accommodate multichannel pipets (at least eight channels) and 96-well plates.
Heavy-duty, one-piece combs eliminate the need for comb assembly. The SuperSafe™ safety lid eliminates the risk of accidental shock. System is supplied with a 0.91m (3') safety power cord, attached.
The buffer chamber features lane visualization strips, which are silkscreened on the platform to provide a clear view of sample wells. Lane markers and fluorescent rulers provide accurate fragment size determination of DNA when compared to known standards. Durable acrylic construction and research-quality components give the buffer chamber a long life. Platinum electrodes and gold-plated banana plugs are corrosion-protected, and electrode placement ensures a uniform electric field.