Chemware® PTFE Boiling Stones, Saint-Gobain Performance Plastics
# de catalogue 26397-103
Fournisseur: Saint Gobain Performance Plastics
Spécifications
- Description:PTFE boiling stones
- Package:450 g
- Supplier No.:D1069103
Spécifications
A propos de cet article
Ultra-pure, virgin PTFE chips.
- promote gentle, efficient boiling by creating many projections on which small bubbles can form
- Unaffected by boiling acids, hydrocarbons, ketones, esters, alcohols, or alkalies in any concentrations
- Suitable for column packing
Ideal for continuous extraction methods, such as EPA SW846.
Packaging: Supplied in a 450 g bottle.
Customers Who Bought This Also Bought