ContactUsLinkComponent
Chemware* PTFE Boiling Stones
Chemware® PTFE Boiling Stones, Saint-Gobain Performance Plastics
# de catalogue 26397-103
Chemware* PTFE Boiling Stones
Chemware® PTFE Boiling Stones, Saint-Gobain Performance Plastics
# de catalogue 26397-103
Spécifications
  • Description:
    PTFE boiling stones
  • Package:
    450 g
  • Supplier No.:
    D1069103
Spécifications
A propos de cet article
Ultra-pure, virgin PTFE chips.
  • promote gentle, efficient boiling by creating many projections on which small bubbles can form
  • Unaffected by boiling acids, hydrocarbons, ketones, esters, alcohols, or alkalies in any concentrations
  • Suitable for column packing
Ideal for continuous extraction methods, such as EPA SW846.
Packaging: Supplied in a 450 g bottle.
Customers Who Bought This Also Bought