Order Entry
ContactUsLinkComponent
Borosilicate Glass Balls
Borosilicate Glass Balls, Walter Stern
# de catalogue 26396-630
Fournisseur:  Walter Stern
Borosilicate Glass Balls
Borosilicate Glass Balls, Walter Stern
# de catalogue 26396-630
Fournisseur:  Walter Stern

Spécifications

  • Ø:
    3 mm
  • Cat. no.:
    26396-630
  • Supplier No.:
    102C

Spécifications

A propos de cet article

Balls are solid borosilicate glass

Diameter and sphericity variation is a maximum ±10% of glass ball diameter. Supplied in one-pound packages.

Customers Who Bought This Also Bought