Accessories for Barnstead B-pure™ Cartridge Water Purification System, Thermo Scientific
Catalog # 26302-618
Supplier: Thermo Fisher Scientific
Specifications
- Description:Pressurized Cartridge
- Cat. no.:26302-618
- Resin Type:Ultrapure, mixed bed
- Supplier No.:D5027