Accessories for Barnstead EASYpure® II Water Purification Systems, Thermo Scientific
Catalog # 26302-465
Supplier: Thermo Fisher Scientific
Specifications
- Description:Hollow Fiber Filter, Sterile, Autoclavable
- Cat. no.:26302-465
- For Use With:All systems
- Supplier No.:FL703X2