ContactUsLinkComponent
VWR* Tri-Pour* Graduated Disposable Beakers
VWR® Tri-Pour® Graduated Disposable Beakers, Polypropylene
Catalog # 25384-158
Supplier:  VWR International
VWR* Tri-Pour* Graduated Disposable Beakers
VWR® Tri-Pour® Graduated Disposable Beakers, Polypropylene
Catalog # 25384-158
Supplier:  VWR International
Specifications
  • Capacity:
    800 ml (27 oz.)
  • Cat. no.:
    25384-158
Specifications
About this item
Transparent polypropylene beaker with three pouring lips for dripless dispensing.
  • Polypropylene construction
  • Transparent
  • Graduated markings
  • Disposable design
Graduated, with accuracy of ±5%. May also be used for measuring. Resistant to strong acids and bases, organic solvents, and commonly used laboratory reagents. Write-on cardboard caps ensure spillproof, leak-resistant closure.
Customers Who Bought This Also Bought