VWR® Tri-Pour® Graduated Disposable Beakers, Polypropylene
# de catalogue 25384-150
Fournisseur: VWR International
Spécifications
- Capacity:50 ml (1.7 oz.)
Spécifications
A propos de cet article
Transparent polypropylene beaker with three pouring lips for dripless dispensing.
- Polypropylene construction
- Transparent
- Graduated markings
- Disposable design
Graduated, with accuracy of ±5%. May also be used for measuring. Resistant to strong acids and bases, organic solvents, and commonly used laboratory reagents. Write-on cardboard caps ensure spillproof, leak-resistant closure.
Customers Who Bought This Also Bought