Chemware® Teflon® Resin Evaporating Dishes, Saint-Gobain Performance Plastics
# de catalogue 25337-059
Fournisseur: Saint Gobain Performance Plastics
Spécifications
- Capacity:100 ml
- Cat. no.:25337-059
- Supplier No.:D1069044
Spécifications
A propos de cet article
With broad pouring spouts and flat bottoms
These resin evaporating dishes, saint-gobain performance plastics are made of PTFE.
Chemically inert to virtually all materials. Heat resistant to 260°C (550°F).
Customers Who Bought This Also Bought