PYREX® Flat-Bottom Evaporating Dishes, Corning
# de catalogue 25315-024
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Capacity:140 ml
- Cat. no.:25315-024
- Diameter ×H:80 x 45 mm
- Supplier No.:3180-80
Spécifications
A propos de cet article
PYREX® borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Dishes with rugged beaded rims and standard medium depth.
Customers Who Bought This Also Bought