Spectra/Por® Closures for Dialysis Membrane Tubing, Spectrum® Laboratories
# de catalogue 25224-159
Fournisseur: Spectrum Laboratories
Small Business Enterprise
Spécifications
- Width:35 mm
- Color:Orange/White
- Description:Set of 10 Standard and 10 Weighted Closures
- Cat. no.:25224-159
- Supplier No.:132751
Spécifications
A propos de cet article
These are used in combination with a standard closure for a vertical use of the dialysis tubing and a magnetic stirrer for continuous stirring of the tubing.
PP, autoclavable closures are lighter than water, normally used in combination with weighted closures. It is only for regenerated cellulose membranes.
Customers Who Bought This Also Bought