Spectra/Por® 1 Dialysis Membranes, MWCO 6000 to 8000, Spectrum® Laboratories
# de catalogue 25218-093
Fournisseur: Spectrum Laboratories
Small Business Enterprise
Spécifications
- Ø:25.5 mm
- Length:30.5 m
- Flat Width:40 mm
- Supplier No.:132660
Spécifications
A propos de cet article
Standard Regenerated Cellulose (RC) is composed of natural cellulose regenerated from cotton linters, therefore these membranes are perfectly suited for a broad range of laboratory dialysis.
The clear and flexible membrane type with a symmetric porosity is economical and sturdy, ideal for desalting, buffer exchanges, macromolecular purifications and other general applications. Possessing a broad range of chemical compatibilities, Standard RC membranes can be used with dilute strong acids and bases, concentrated weak acids and bases, most alcohols, and some mild or dilute organic solvents, including DMSO.
Customers Who Bought This Also Bought