Nalgene® CryoSleeve, Polyvinyl Chloride, Thermo Scientific
# de catalogue 24270-024
Fournisseur: Thermo Fisher Scientific
Spécifications
- Length:273 mm (103/4")
- Description:CryoSleeve
- Cat. no.:24270-024
- Supplier No.:5016-0001
Spécifications
A propos de cet article
Clear plastic sleeve encloses a NALGENE® CryoCane™ for extra security during handling and storage
Direct replacement for cardboard sleeve. Fits over most canes. Allows quick location of empty spaces in the cane and easy identification of a particular vial or vials without removing the sleeve. Will not become brittle during freezing or thawing.
Customers Who Bought This Also Bought