Disposable Scalpels, Sterile, Integra™ Miltex® | Avantor
Disposable Scalpels, Sterile, Integra™ Miltex®
#11
Catalog # 21909-656
Supplier:   Miltex
Fournisseur #:  4-411
Disposable Scalpels, Sterile, Integra™ Miltex®
Catalog # 21909-656
Supplier:  Miltex
Disposable Scalpels, Sterile, Integra™ Miltex®
Disposable Scalpels, Sterile, Integra™ Miltex®
#11
Price
$95.29
Log in pour la disponibilité
Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Scalpels feature plastic handles and razor-sharp, stainless steel blades for superior cutting and clean incisions
Size is clearly marked on each blade. Safety sleeve protects both user and blade. Individually packaged in sealed, peel-apart pouches. Five-year shelf life.
Documents
Spécifications
  • Blade No.:
    #11