ContactUsLinkComponent
Scalpel Blades, Sterile
Disposable Scalpel Blades, Sterile, Integra™ Miltex®
# de catalogue 21909-628
Fournisseur:  Miltex
Scalpel Blades, Sterile
Disposable Scalpel Blades, Sterile, Integra™ Miltex®
# de catalogue 21909-628
Fournisseur:  Miltex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Carbon Steel Blade
  • Cat. no.:
    21909-628
  • Blade Size:
    #23
  • Supplier No.:
    4-123
Spécifications
A propos de cet article
Carbon steel and stainless steel surgical blades feature razor-sharp cutting edges for superior cutting and clean incisions
Blades are for use with No. 3 or No. 4 scalpel handles. No. 3 scalpel handles accommodate blade numbers 10 through 15C. No. 4 scalpel handles accommodate blade numbers 20 through 25. Size is clearly marked on each blade. Individually packaged in sealed, peel-apart pouches. Five-year shelf life.
Customers Who Bought This Also Bought