Specifications
- Ø:5.0 mm
- Cat. no.:21909-142
- Shape:Straight
- Supplier No.:33-35
Specifications
About this item
These biopsy punches offer the convenience of single-use disposability without sacrificing quality, feel, or performance
All punches feature seamless, stainless-steel, razor-sharp edges to eliminate torn or pulled biopsies. Ribbed handles provide comfort and control. Punches are color-coded with sizes clearly marked. Sterile, five-year shelf life.
Plunger-system models are available in five sizes and offer the added benefit of effortless ejection of lodged speciments. Non-plunger-system models are available in ten sizes for a wide range of procedures.