Spécifications
- Size:11 kg (25 lbs.)
- Cat. no.:21837-151
- Packaging:Box
- pH:9.5
- Supplier No.:1325
Spécifications
À propos de cet article
Tergazyme is a concentrated, anionic detergent with protease enzyme for manual and ultrasonic cleaning. Excellent for removal of proteinaceous soils, tissue, blood and body fluids from glassware, metals, plastic, ceramic, porcelain, rubber and fiberglass with no interfering residues. Ideal as a cleaning agent in Reverse Osmosis and Ultra-Filtration Systems.
- Biodegradable and readily disposable
- Replaces corrosive acids and hazardous solvents
- Protease enzyme removes proteinaceous soils, tissue, blood and body fluids
- Free rinsing to give you reliable results and no interfering residues
- Use to pass your cleaning validation tests for lab accreditation and plant inspection approval
It can be used to clean hospital instruments, dairy equipment, laboratory ware, reverse osmosis and ultrafiltration membranes and units, sampling apparatus, pharmaceutical apparatus, cosmetics manufacturing equipment, tubing, pipes, optical parts, process equipment, industrial parts, desalination plants, tanks and reactors. Dilute 1:100.
And it can be used to remove soil, grit, grime, blood, tissue, grease, fats, oils, proteinaceous soils, dairy proteins, particulates, solvents and bioreactor residue.
Corrosion inhibited formulation recommended for glass, metal, stainless steel, porcelain, ceramic, plastic, rubber and fiberglass. Can be used on soft metals such as copper, aluminum, zinc and magnesium if rinsed promptly. Corrosion testing may be advisable.
Cleaning method: Soak, brush, sponge, cloth, ultrasonic, flow through clean-inplace.