Spécifications
- Size:56.8 L (15 gal.)
- Cat. no.:21834-116
- pH:7
- Supplier No.:1915
- Packing Type:Drum
Spécifications
A propos de cet article
Luminox is a neutral pH detergent for use in manual, ultrasonic, and machine cleaning. Safe-handling detergent used for non-corrosive cleaning without chelation or alkalinity waste treatment problems. Recommended for residue free cleaning of aluminum and soft metal, medical devices, pharmaceutical equipment, alkaline sensitive labware, and medical instruments in aluminum trays. Free rinsing, eliminating interfering residues.
- Low foaming for use in mechanical and spray washers
- No chelation or alkalinity waste treatment problems
- Concentrated for economical use
- Biodegradable and phosphate free
- Dilute 3:100
For removing oils, dirt, grime, adhesives and uncured epoxy, particulates, residues, fluxes, chemical solvents, machining oils.
Cleaning method: Soak tanks, manual cleaning, ultrasonic tanks, mechanical washers, washer/sanitizers, spray washers, CIP systems.
Corrosion inhibited formulation recommended for glass, ferrous metals, non-ferrous metals, aluminum, copper, zinc, and other soft metals, ceramic, plastic, circuit boards and electronic components.