ContactUsLinkComponent
Luminox* Low-Foaming Neutral Cleaner
Luminox® Low-Foaming Neutral Cleaners
# de catalogue 21834-116
Fournisseur:  Alconox
Luminox* Low-Foaming Neutral Cleaner
Luminox® Low-Foaming Neutral Cleaners
# de catalogue 21834-116
Fournisseur:  Alconox
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    56.8 L (15 gal.)
  • Cat. no.:
    21834-116
  • pH:
    7
  • Supplier No.:
    1915
  • Packing Type:
    Drum
Spécifications
A propos de cet article
Luminox is a neutral pH detergent for use in manual, ultrasonic, and machine cleaning. Safe-handling detergent used for non-corrosive cleaning without chelation or alkalinity waste treatment problems. Recommended for residue free cleaning of aluminum and soft metal, medical devices, pharmaceutical equipment, alkaline sensitive labware, and medical instruments in aluminum trays. Free rinsing, eliminating interfering residues.
  • Low foaming for use in mechanical and spray washers
  • No chelation or alkalinity waste treatment problems
  • Concentrated for economical use
  • Biodegradable and phosphate free
  • Dilute 3:100
For removing oils, dirt, grime, adhesives and uncured epoxy, particulates, residues, fluxes, chemical solvents, machining oils.
Cleaning method: Soak tanks, manual cleaning, ultrasonic tanks, mechanical washers, washer/sanitizers, spray washers, CIP systems.
Corrosion inhibited formulation recommended for glass, ferrous metals, non-ferrous metals, aluminum, copper, zinc, and other soft metals, ceramic, plastic, circuit boards and electronic components.
Certifications: FDA certified.
Customers Who Bought This Also Bought