Glass Wool and Extractor, Ohio Valley
# de catalogue 21507-980
Fournisseur: Ohio Valley
Small Business Enterprise
Spécifications
- Description:DMCS (Silanized) Treated Glass Wool, 50 g
- Cat. no.:21507-980
- Supplier No.:3352
Spécifications
A propos de cet article
Borosilicate glass wool, either untreated or treated with a dimethyldichlorosilane solution to render the wool inert for use in gas chromatography.
Fiber Diameter: 8 micron
Glass wool extractor: Insert extractor in column, twist slightly to hook glass wool and pull.
Customers Who Bought This Also Bought