ContactUsLinkComponent
Tube, Shortwave
Accessories for UVP Blak-Ray® and Sterilizing UV Bench/Display Lamps, Analytik Jena
# de catalogue 21476-009
Fournisseur:  Analytik Jena US
Small Business Enterprise
Accessories for UVP Blak-Ray® and Sterilizing UV Bench/Display Lamps, Analytik Jena
# de catalogue 21476-009
Fournisseur:  Analytik Jena US
Small Business Enterprise
Spécifications
  • Description:
    Replacement Tube
  • For Model:
    XX-15S
  • Wavelength:
    254 nm
  • Supplier No.:
    34-0008-01