Accessories for UVP Blak-Ray® and Sterilizing UV Bench/Display Lamps, Analytik Jena
# de catalogue 21476-009
Fournisseur: Analytik Jena US
Small Business Enterprise
Spécifications
- Description:Replacement Tube
- For Model:XX-15S
- Wavelength:254 nm
- Supplier No.:34-0008-01