Accessories for UVP EL Series Ultraviolet Lamps, Analytik Jena
# de catalogue 21474-965
Fournisseur: Analytik Jena US
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Description:Replacement Tube, Mid-Range, 302 nm, 8 W
- Cat. no.:21474-965
- For Model:UVM-18, UVM-28, C65
- Supplier No.:34-0042-01
Spécifications
A propos de cet article
Customers Who Bought This Also Bought