Order Entry
ContactUsLinkComponent
Conical-Bottom Centrifuge Tube
Disposable Plain Centrifuge Tubes, Glass, Kimble Chase
Catalog # 21020-695
Conical-Bottom Centrifuge Tube
Disposable Plain Centrifuge Tubes, Glass, Kimble Chase
Catalog # 21020-695

Specifications

  • Cap size:
    24-400
  • Ø×L:
    29 x 137 mm
  • Volume:
    50 ml
  • Cat. no.:
    21020-695
  • Maximum RCF:
    2980×g
  • Supplier No.:
    73785-50

Specifications

About this item

These borosilicate glass tubes have a conical bottom.

  • 50 ml tube has a bullet nose for extra strength

Caps are phenolic screw type with a PTFE-faced rubber liner.

Certifications: Meet Type I, A, or B glass requirements per ASTM E438.
Ordering information: Supplied in shrink-wrapped trays without caps. Caps are ordered separately.
Customers Who Bought This Also Bought