Corning® Centrifuge Tubes, Sterile, Polypropylene, Corning
# de catalogue 21008-771
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Volume:250 ml
- Maximum RCF:6000×g
- Supplier No.:430776
Spécifications
A propos de cet article
PP, with HDPE plug seal cap, sterile. Polyetherimide cushions.
- Moulded graduations, conical bottom
- Tubes are sterile and certified non pyrogenic
- Support cushions must be used unless the rotor has appropriately shaped V-bottom holders
Ideal for applications requiring large volume centrifugation.
They are perfect for large volume centrifugation applications. They incorporate plug seal caps and have molded graduations.
Ordering information: Non sterile cushion sold separately.
Packaging: Bulk, 6/bag.
Customers Who Bought This Also Bought