VWR® Microcentrifuge Tube Rack, PP
Catalogue # 20901-675
Fournisseur: VWR International
Spécifications
- Dimensions:75 x 210 x 35 mm (2.9 x 8.3 x 1.4")
- Cat. no.:20901-675
Spécifications
À propos de cet article
Each polypropylene microcentrifuge tube rack stores 20 standard 1.5–2.0 mL microcentrifuge tubes in clearly numbered cavities.
- Numbered cavities for quick identification
- One-handed transportation
- Autoclavable
- Polypropylene construction
Easily transportable with one hand. Made of autoclavable polypropylene. Non-skid rubber feet.
Frequently Bought Together