Deflected Point Septum Penetration Needles, Cadence Science®
# de catalogue 20068-586
Fournisseur: Cadence Science
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Length:152.4 mm (6.00")
- Description:Std. Plated Hub with 304 SS Cannula
- Gauge:20
- Supplier No.:4186
Spécifications
A propos de cet article
The deflected needle features point positioning on the axis of the needle lumen
This improved point geometry provides easier penetration and minimizes coring and tracking (particularly with finer gauge needles)
Ordering information: Non-sterile. Packaged 12 needles per box.
Customers Who Bought This Also Bought