To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Standard Spore Strips
- Application:Steam Sterilization
- Organism:Geobacillus stearothermophilus
- Spore Population Per Strip:10⁵/strip
- Supplier No.:STS-05
Spécifications
A propos de cet article
Biological sterilization indicators are used to perform sterilization cycle validation.
When stored under labeled conditions, all spore strips listed have a shelf life of 18 months, except mini spore strips 19872-014, which have a shelf life of 24 months.
They are created by inoculating spore suspensions of certified population and resistance onto paper strips. Standard spore strips and combination spore strips are 7x38 mm. Mini spore strips are 2x10 mm in size and will fit into small areas of a device where a standard-sized spore strip cannot be used, such as within a syringe barrel, inside tubing, or under a cap.