NALGENE® Polypropylene Screw Caps, Thermo Scientific
# de catalogue 18060-136
Fournisseur: Thermo Fisher Scientific
Spécifications
- Cap Size:24–415
- Color:Red
- Cat. no.:18060-136
- Supplier No.:362150-5240
Spécifications
A propos de cet article
Linerless caps are for use only with NALGENE® narrow- and wide-mouth bottles and Erlenmeyer flasks
Note: Before autoclaving, set cap on top of container without engaging threads.
Autoclavable.
Ordering information: Bulk packed.
Customers Who Bought This Also Bought