ContactUsLinkComponent
Nalgene Polypropylene Screw Caps, Thermo Scientific
NALGENE® Polypropylene Screw Caps, Thermo Scientific
# de catalogue 18060-136
Nalgene Polypropylene Screw Caps, Thermo Scientific
NALGENE® Polypropylene Screw Caps, Thermo Scientific
# de catalogue 18060-136
Spécifications
  • Cap Size:
    24–415
  • Color:
    Red
  • Cat. no.:
    18060-136
  • Supplier No.:
    362150-5240
Spécifications
A propos de cet article
Linerless caps are for use only with NALGENE® narrow- and wide-mouth bottles and Erlenmeyer flasks
Note: Before autoclaving, set cap on top of container without engaging threads.
Autoclavable.
Ordering information: Bulk packed.
Customers Who Bought This Also Bought