Nalgene® Unitary™ Safety Wash Bottles, Thermo Scientific
Catalog # 16651-961
Supplier: Thermo Fisher Scientific
Specifications
- Capacity:500 ml (16.9 oz.)
- Cat. no.:16651-961
- Supplier No.:DS2408-0500
Specifications
About this item
Nalgene® Unitary™ Safety Wash Bottles are molded in one piece which eliminates possible contamination from a delivery tube and offers safety from chemical splashes when removing the closure.
The bottles also feature a leakproof linerless polypropylene closure, and are red for easy identification.
Customers Who Bought This Also Bought