ContactUsLinkComponent
Biohazard Wipers
Biohazard Wipes and Liners, Current Technologies
# de catalogue 16200-212
Fournisseur:  Current Technologies
Minority or Woman-Owned Business Enterprise
Biohazard Wipers
Biohazard Wipes and Liners, Current Technologies
# de catalogue 16200-212
Fournisseur:  Current Technologies
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Dimensions:
    7.6 x 7.6 cm (3 x 3")
  • Description:
    Super Absorbency Biohazard Wipes
  • Supplier No.:
    BH91033
Spécifications
A propos de cet article
Low-linting wipes and liners are specifically designed to protect workers and labs from contamination from blood, body fluids, tissue, specimens plus chemicals.
Wipes and liners are available in several sizes and in large rolls for cutting to size. Wipes are also available in standard absorbency, which can absorb 9× its own weight, heavy absorbency, which can absorb 12× its weight and super absorbency, which can absorb 25× its weight.
Specifically sized dispensers are constructed of clear acrylic.
Absorbent white side soaks up blood, tissue, specimen, body fluids, and chemicals. Bright orange barrier side, made of coextruded plastic, blocks soak-through to gloves, hands and counters, limiting the spread of contamination. Wipes are impermeable to strong solvents including xylene and xylene substitutes, acetone, ketones, alcohols, and formalin. Use to pop caps, wipe pipets and probes, handle specimens and line benches, counters and trays, clean up spills.
Ordering information: Bio-Screen wipes and liners are non-sterile and are not for use on patients.
Customers Who Bought This Also Bought