Spécifications
- Dimensions:10.2 x 10.2 cm (4 x 4")
- Description:Super Absorbency Biohazard Wipes
- Supplier No.:BH90044
Spécifications
A propos de cet article
Low-linting wipes and liners are specifically designed to protect workers and labs from contamination from blood, body fluids, tissue, specimens plus chemicals.
Wipes and liners are available in several sizes and in large rolls for cutting to size. Wipes are also available in standard absorbency, which can absorb 9× its own weight, heavy absorbency, which can absorb 12× its weight and super absorbency, which can absorb 25× its weight.
Specifically sized dispensers are constructed of clear acrylic.
Absorbent white side soaks up blood, tissue, specimen, body fluids, and chemicals. Bright orange barrier side, made of coextruded plastic, blocks soak-through to gloves, hands and counters, limiting the spread of contamination. Wipes are impermeable to strong solvents including xylene and xylene substitutes, acetone, ketones, alcohols, and formalin. Use to pop caps, wipe pipets and probes, handle specimens and line benches, counters and trays, clean up spills.