ContactUsLinkComponent
KIM-KAP* Polypropylene Cap
KIM-KAP® Caps, Disposable, Polypropylene, Kimble Chase
# de catalogue 16199-010
KIM-KAP* Polypropylene Cap
KIM-KAP® Caps, Disposable, Polypropylene, Kimble Chase
# de catalogue 16199-010
Spécifications
  • Color:
    Blue
  • Cat. no.:
    16199-010
  • For Tube O.D.:
    16 mm
  • Supplier No.:
    73665-16
Spécifications
A propos de cet article
These KIM caps are designed to fit culture tubes with outer diameters ranging from 13 mm to 38 mm.
  • Autoclavable
  • Color coded
Customers Who Bought This Also Bought