Nalgene® Economy Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Catalogue # 16172-164
Fournisseur: Thermo Fisher Scientific
Spécifications
- Cap Size:38-415
- Capacity:125 ml (4.2 oz.)
- Color:Natural
- Cat. no.:16172-164
- Bottle Style:Wide mouth
- Supplier No.:2189-0004
Spécifications
À propos de cet article
Economical, single-use, break-resistant bottles feature thinner walls and are more flexible than original Nalgene® bottles.
- Excellent chemical resistance to most acids, bases, and alcohols
- Available in natural and amber finishes
- Guaranteed leakproof
Ideal for short-term, general-purpose laboratory and field applications, such as collecting samples, or for storing working solutions like buffers, bench reagents, and diluted chemical aliquots. Amber bottles reduce UV light transmission for protection of light-sensitive liquids.
Certifications: Amber bottles comply with U.S. Pharmacopoeia 23 requirements for maximum light transmission.
Delivery information: Supplied with polypropylene screw closures.
Caution: Not recommended for hazardous and critical material containment.
Frequently Bought Together