Nalgene® Economy Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Catalog # 16172-160
Supplier: Thermo Fisher Scientific
Specifications
- Cap size:28-415
- Capacity:30 ml (1 oz.)
- Color:Natural
- Environmentally Preferable:Product Transparency and Disclosure
- Environmental certification:ACT Label
- Cat. no.:16172-160
- Bottle Style:Wide mouth
- Supplier No.:2189-0001
Specifications
About this item
Economical, single-use, break-resistant bottles feature thinner walls and are more flexible than original Nalgene® bottles.
- Excellent chemical resistance to most acids, bases, and alcohols
- Available in natural and amber finishes
- Guaranteed leakproof
Ideal for short-term, general-purpose laboratory and field applications, such as collecting samples, or for storing working solutions like buffers, bench reagents, and diluted chemical aliquots. Amber bottles reduce UV light transmission for protection of light-sensitive liquids.
Certifications: Amber bottles comply with U.S. Pharmacopoeia 23 requirements for maximum light transmission.
Delivery: Supplied with polypropylene screw closures.
Caution: Not recommended for hazardous and critical material containment.
Customers Who Bought This Also Bought