ContactUsLinkComponent
Aluminum Seal
Aluminum Seals, Center Disc Tear-Out, Unlined, WHEATON®
Catalog # 16171-828
Small Business Enterprise
Aluminum Seal
Aluminum Seals, Center Disc Tear-Out, Unlined, WHEATON®
Catalog # 16171-828
Small Business Enterprise
Specifications
  • Material:
    Green Aluminum
  • Cat. no.:
    16171-828
  • For O.D.:
    20 mm
  • Supplier No.:
    224183-07
Specifications
About this item
One-piece center disc tears out, exposing top of seal to syringe
Shelf packs of 100.
Customers Who Bought This Also Bought