Aluminum Seals, Center Disc Tear-Out, Unlined, WHEATON®
Catalog # 16171-828
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Material:Green Aluminum
- Cat. no.:16171-828
- For O.D.:20 mm
- Supplier No.:224183-07
Specifications
About this item
One-piece center disc tears out, exposing top of seal to syringe
Shelf packs of 100.
Customers Who Bought This Also Bought