Straight Plug Rubber Stoppers, Wheaton
# de catalogue 16171-680
Fournisseur: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Spécifications
- Description:Straight plug rubber stoppers, durameter 45
- Material:Natural red rubber
- Fits:Mouth with ID×OD 13×20 mm
- Supplier No.:224100-172
Spécifications
A propos de cet article
Wheaton straight plug stoppers provide maximum sealing for vials with a straight-wall glass finish
Stoppers are autoclavable.
Stoppers are available in a variety of sizes and rubber formulations . Straight plug stoppers are manufactured from the highest quality raw materials and are manufactured to yield exceptional dimensional stability.
Customers Who Bought This Also Bought