Serum Bottles, Borosilicate Glass, WHEATON®, DWK Life Sciences
# de catalogue 16171-433
Fournisseur: DWK LIFE SCIENCES S. DE R.L. DE MX
Spécifications
- Capacity:10 ml (0.34 oz.)
- Material:Glass, amber
- Diameter ×H:25 x 54 mm
- Supplier No.:223761
Spécifications
A propos de cet article
Serum bottles are ideal for long and short term sample storage, lyophilization, and as vaccine/injectable drug containers.
- Bottles fit most lyophilization applications
Clear bottles are manufactured from Wheaton 400 borosilicate molded glass, and conform to USP Type I requirements. Amber bottles are manufactured from Wheaton 500 borosilicate glass, and conform to USP Type I requirements. All bottles feature a 13Int.Ø×20Ext.Ø mm mouth opening, with the exception of 89422-806 which is 7Int.Ø×13Ext.Ø mm.
Ordering information: Bottles are packaged in shrink-wrapped, partitioned modules to reduce breakage.
Customers Who Bought This Also Bought