Order Entry
ContactUsLinkComponent
Media Bottles, Graduated
Media Bottles, Graduated, WHEATON®, DWK Life Sciences
# de catalogue 16159-980
Small Business Enterprise
Media Bottles, Graduated
Media Bottles, Graduated, WHEATON®, DWK Life Sciences
# de catalogue 16159-980
Small Business Enterprise

Spécifications

  • Cap Size:
    33-430
  • Capacity:
    500 ml (16.9 oz.)
  • Graduation range:
    100–400 mL
  • Description:
    With Rubber-Lined Cap
  • Supplier No.:
    219759

Spécifications

A propos de cet article

Borosilicate glass bottles have approximate graduations at 50mL intervals and a large writing area.

  • Clear
  • Type l borosilicate glass bottles
  • Fluoropolymer resin-lined caps and rubber-lined caps are autoclavable

Large opening and no-drip pour ring make bottles easy to clean. Choose from bottles without caps, with fluoropolymer resin-lined caps, polyethylene-lined caps, or rubber-lined caps.

Customers Who Bought This Also Bought