ContactUsLinkComponent
Corning* GL45 Red High Temperature (180 degree C) PBT Screw Cap with PTFE faced silicone liner
Accessories for PYREX® Media Bottles, Graduated, Corning
# de catalogue 16157-464
Fournisseur:  Corning
Product available on GSA Advantage®
Corning* GL45 Red High Temperature (180 degree C) PBT Screw Cap with PTFE faced silicone liner
Accessories for PYREX® Media Bottles, Graduated, Corning
# de catalogue 16157-464
Fournisseur:  Corning
Product available on GSA Advantage®
Spécifications
  • Cap Size:
    GL45
  • Description:
    High-Temperature PBT Cap, Red
  • Cat. no.:
    16157-464
  • Supplier No.:
    1395-45HTC