ContactUsLinkComponent
Colored Starline Caps For Starline Narrow Mouth Containers, White
WHEATON® Polypropylene Screw Cap, Linerless, DWK Life Sciences
# de catalogue 16153-475
Small Business Enterprise
Colored Starline Caps For Starline Narrow Mouth Containers, White
WHEATON® Polypropylene Screw Cap, Linerless, DWK Life Sciences
# de catalogue 16153-475
Small Business Enterprise
Spécifications
  • Color:
    White
  • Supplier No.:
    239506-02
Spécifications
A propos de cet article
A variety of polypropylene caps are offered separately for Wheaton Leak Resistant Narrow Mouth Bottles
Caps can be autoclaved for 15 minutes at 121ºC at 15 psi. Polypropylene caps are available in natural, white, red, blue and green
Customers Who Bought This Also Bought