KIMBLE® RAY-SORB® Amber Erlenmeyer Flask, DWK Life Sciences
# de catalogue 16143-156
Fournisseur: DWK LIFE SCIENCES S. DE R.L. DE MX
Spécifications
- Capacity:125 ml
- Cat. no.:16143-156
- Stopper No.:22
- Supplier No.:26610-125
Spécifications
A propos de cet article
These flasks are RAY-SORB® processed to protect solutions sensitive to light from shorter wavelengths.
No capacity scale provided. Designed from ASTM Specification E1404, Type II, Class 2 requirements.
Customers Who Bought This Also Bought