ContactUsLinkComponent
Flasks Erlenmeyer RAY-SORB Color-Coded PTFE [standard taper] Stopper
KIMBLE® RAY-SORB® Amber Erlenmeyer Flask, DWK Life Sciences
# de catalogue 16143-156
Flasks Erlenmeyer RAY-SORB Color-Coded PTFE [standard taper] Stopper
KIMBLE® RAY-SORB® Amber Erlenmeyer Flask, DWK Life Sciences
# de catalogue 16143-156
Spécifications
  • Capacity:
    125 ml
  • Cat. no.:
    16143-156
  • Stopper No.:
    22
  • Supplier No.:
    26610-125
Spécifications
A propos de cet article
These flasks are RAY-SORB® processed to protect solutions sensitive to light from shorter wavelengths.
No capacity scale provided. Designed from ASTM Specification E1404, Type II, Class 2 requirements.
Customers Who Bought This Also Bought