Nalgene® High-Density Polyethylene Economy Bottles, Bulk Pack, Wide Mouth, Thermo Scientific
# de catalogue 16060-008
Fournisseur: Thermo Fisher Scientific
Spécifications
- Cap Size:38 mm
- Capacity:125 ml (4.2 oz.)
- Ø:50.5 mm
- Cat. no.:16060-008
- Supplier No.:332189-0004
- Height with Cap:100.8 mm
- Height without Cap:98.3 mm
Spécifications
A propos de cet article
Nalgene® Economy Bottles are lightweight, more flexible versions of the original Nalgene Lab Quality Bottles for less rigorous lab use.
- Priced below low-quality plastic or glass bottle brands
- Economical, quality choice for lighter-duty lab use
These Nalgene® Economy Bottles feature excellent impact and chemical resistance and clarity, include a polypropylene closure, and are made from natural, translucent high-density polyethylene. Like most Nalgene bottles, the Economy Bottles are guaranteed leakproof and are made in our ISO-certified USA manufacturing facility.
These bottles are suitable for light-duty, general purpose laboratory applications such as containing samples and daily working solutions like buffers and bench reagents.
Ordering information: Packaged in bulk quantities. Direct UV exposure should be avoided.
Customers Who Bought This Also Bought