Nalgene® IP2 Narrow Mouth Bottles, Natural HDPE, with PP Closure, Bulk Pack, Thermo Scientific
# de catalogue 16059-716
Fournisseur: Thermo Fisher Scientific
Spécifications
- Cap Size:20 mm
- Capacity:60 ml (2 oz.)
- Cat. no.:16059-716
- Neck I.D.:14 mm
- Outer Diameter:39 mm
- Supplier No.:312099-0002
- Height with Cap:84 mm
Spécifications
A propos de cet article
Nalgene® IP2 high density polyethylene narrow mouth bottles are recommended for storage and shipment of liquid reagents.
These bottles come with polypropylene closures and are recommended for customers involved in designing, assembling and certifying their own combination packaging. The bottles are safe, economical, and comply with the IATA and ICAO regulations.
Packaging: Bottles and closures are bulk packed in separate polybags within the same carton.
Customers Who Bought This Also Bought