ContactUsLinkComponent
VWR Pre-Filled Formalin Container
VWR® Prefilled Histology Containers, 10% Neutral Buffered Formalin (NBF)
# de catalogue 16004-112
Fournisseur:  VWR International
Product available on GSA Advantage®
VWR Pre-Filled Formalin Container
VWR® Prefilled Histology Containers, 10% Neutral Buffered Formalin (NBF)
# de catalogue 16004-112
Fournisseur:  VWR International
Product available on GSA Advantage®
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Capacity:
    20 ml
  • Description:
    Prefilled 10% Neutral Buffered Formalin
Spécifications
A propos de cet article
Containers are half-filled with 10% neutral buffered formalin, for the collection and transport of tissue samples.
Prefilled polypropylene containers feature polyethylene screw caps with dual thread closures and a unique integrated leak-resistant seal. Bulk formalin is packaged in polyethylene Cubitainer® containers.
  • High clarity virgin polypropylene container construction
  • Polyethylene screw caps included
  • 20–120 mL containers are 95kPa compliant
Containers are half-filled with 10% neutral buffered formalin, for the collection and transport of tissue samples.
Each container is made of high clarity virgin polypropylene and includes a polyethylene screw cap
Each container is made of high clarity virgin polypropylene and includes a polyethylene screw cap
Small-sized prefilled formalin containers (20 mL and 40 mL) feature a click-top closure. The first click assures users the seal is fully engaged. The second click ensures 95kPa compliance. This click-top design prevents leaking formalin containers and ensures safe specimen transport.
Containers sized 20 mL through 120 mL are packaged in transport packs with handles. This packaging design allows for easy distribution to other labs and doctors’ offices. The inner packaging is made of chipboard material, and is safe for operating room use.
Caps feature dual thread closures and a leak-resistant seal to protect specimens. All containers sized 20 mL through 120 mL are 95kPa compliant.
Customers Who Bought This Also Bought