ContactUsLinkComponent
MEMBRANE 7X85CM45NTRCEL PK10
Transfer Membranes, NuSep
# de catalogue 16003-904
Fournisseur:  NuSep
Transfer Membranes, NuSep
# de catalogue 16003-904
Fournisseur:  NuSep
Spécifications
  • Description:
    Transfer membrane, Nitrocellulose, sheets, pore size 0.45 µm
  • Cat. no.:
    16003-904
  • Supplier No.:
    RB-700
Spécifications
A propos de cet article
Transfer membranes are 7x8.5cm (23/4x35/16").
Customers Who Bought This Also Bought