Transfer Membranes, NuSep
# de catalogue 16003-904
Fournisseur: NuSep
Spécifications
- Description:Transfer membrane, Nitrocellulose, sheets, pore size 0.45 µm
- Cat. no.:16003-904
- Supplier No.:RB-700
Spécifications
A propos de cet article
Transfer membranes are 7x8.5cm (23/4x35/16").
Customers Who Bought This Also Bought