Reaction Vessel Lids, 6-Necks, Chemglass
# de catalogue 16002-864
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Description:Reaction Kettle Lid, 6-neck, 200 mm
- Cat. no.:16002-864
- Supplier No.:CG-1946-C-10
Spécifications
A propos de cet article
Six-neck lid for 200 mm flanged vessel.
Lid has 45/50 center neck, four 45/50 side necks and a 60 mm angled flange for powder addition.
Customers Who Bought This Also Bought