MPC Series 1/4" Flow, Quick Disconnect Coupling for Bio and Pharma, Colder Products Company
Catalogue # 16001-606
Fournisseur: Colder Products
Spécifications
- Description:Polycarbonate Sealing Cap
- Cat. no.:16001-606
- Supplier No.:MPC32003
Spécifications
À propos de cet article
These 1/4" flow quick disconnect couplings are manufactured specifically for the bio and pharma markets
Materials of construction include animal free (BSE/TSE) USP class 6 polysulfone and polycarbonate, Buna - n and platinum cured USP class 6 silicone oring seals.
USP Class 6 and animal free (BSE/TSE) materials are appropriate for the Bio, Pharma and Life Sciences with single use dispoability in mind.
Certifications: USP Class VI, BSE/TSE free materials (ADCF)
Ordering information: Accessories not included
Frequently Bought Together