ContactUsLinkComponent
Plastic Narrow Mouth Commercial Cylinders, With White PP Sturdeeseal P/E Foam Lined Caps Attached
Cylinder Bottles, Natural High-Density Polyethylene, Wide Mouth, Qorpak®
# de catalogue 15712-224
Fournisseur:  Qorpak
Small Business Enterprise
Plastic Narrow Mouth Commercial Cylinders, With White PP Sturdeeseal P/E Foam Lined Caps Attached
Cylinder Bottles, Natural High-Density Polyethylene, Wide Mouth, Qorpak®
# de catalogue 15712-224
Fournisseur:  Qorpak
Small Business Enterprise
Spécifications
  • Cap Size:
    38-400
  • Cap Type:
    White Polypropylene F217 Lined
  • Capacity:
    355 ml (12 oz.)
  • Cat. no.:
    15712-224
  • Supplier No.:
    PLC-03455
Spécifications
A propos de cet article
Wide mouth cylinder bottles offer all the benefits of the cylinder shape and HDPE properties as our commercial cylinders but with the added advantage of a wider opening.
Customers Who Bought This Also Bought