Spécifications
- Capacity:90 ml (3 oz.)
- Ø:48 mm (17/8")
Spécifications
A propos de cet article
Sterile, straight-sided specimen containers are made of break-resistant polypropylene.
- Containers supplied sterile
- Polypropylene container construction
- Polyethylene seals provide excellent leak resistance
Sterile, straight-sided specimen containers are made of break-resistant polypropylene.
The blue polyethylene closure seals with an audible click, assuring superior leak resistance. Closures are tamper-sealed.