ContactUsLinkComponent
VWR* Specimen Container
VWR® ClikSeal Specimen Containers
# de catalogue 15704-002
Fournisseur:  VWR International
VWR* Specimen Container
VWR® ClikSeal Specimen Containers
# de catalogue 15704-002
Fournisseur:  VWR International
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Capacity:
    90 ml (3 oz.)
  • Ø:
    48 mm (17/8")
Spécifications
A propos de cet article
Sterile, straight-sided specimen containers are made of break-resistant polypropylene.
  • Containers supplied sterile
  • Polypropylene container construction
  • Polyethylene seals provide excellent leak resistance
Sterile, straight-sided specimen containers are made of break-resistant polypropylene.
The blue polyethylene closure seals with an audible click, assuring superior leak resistance. Closures are tamper-sealed.
Customers Who Bought This Also Bought