Specifications
- Description:VWR® scientific calculator
- Dimensions:7.6×12.7×1.3 cm (3×5×0.5")
- Cat. no.:15551-112
Specifications
About this item
This scientific calculator performs 99% of routine and not-so-routine lab calculations, and provides instant and accurate answers for a variety of conversions, scientific functions, statistical operations, all arithmetic calculations, and other lab constants.
- Capable of performing over 50 unique functions
- Useful for a range of subjects
- Memory stores data even when unit is off
- Easy-to-carry pocket-sized design
Performs over 50 functions, including science, math, common/hyperbolic trigonometric, logs/natural logs, powers, roots, reciprocals, factorials, percentages, hexadecimals, random number generation, and degree/decimal conversion. Statistical features include standard deviation, mean, sum of squares, sum of data, and much more. Three-key memory and “Forever Memory” store data when the unit is off. Automatic power-down function conserves battery life. Answers are shown on a 10-digit numerical, 2-digit exponent display. This easy-to-carry pocket-sized calculator features raised keys and a large, bright, vivid, 0.6 cm (1/4") high easy-to-read LCD screen.