Spécifications
- Capacity:2 ml (0.06 oz.)
- Description:Sample Cups for Technicon and Other Autoanalyzers
- Cat. no.:15070-290
Spécifications
A propos de cet article
These transparent polystyrene sample cups are resistant to most dilute aqueous solutions.
- Compatible with most autoanalyzers
- Wide base prevents spillage
These transparent polystyrene sample cups are resistant to most dilute aqueous solutions.
Cups are available for use with Technicon®, Abbott®, Hitachi®, and most other autoanalyzers
Autoanalyzer sample cups are not autoclavable.
The wide-base design prevents tipping and spilling. The conical bottom is standard and recommended for microsampling.